Learn More
Cayman Chemical SETD3 MoncLNal ANTIBDYCl 100ug
SDP
Catalog No. 502938297
Encompass_Preferred
Immunogen: Full length recombinant human SETD3 protein • Host: Mouse • Applications: ELISA, WB
Catalog No. 50-293-8297 Supplier Cayman Chemical Supplier No. 29208100 UG
Immunogen: Full length recombinant human SETD3 protein • Host: Mouse • Applications: ELISA, WB