Learn More
Aviva Systems Biology Corp SETD4 Antibody - C-terminal region (ARP34563_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34563_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SETD4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human SETD4. Alternate names: C21orf18, C21orf27ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-55453