Learn More
Aviva Systems Biology Corp Setd5 Antibody - N-terminal region (ARP49310_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49310_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Setd5. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Rat. Alternate names: RGD1310433ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-411