Learn More
Aviva Systems Biology Corp SETD6 Antibody - middle region (ARP51243_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP51243_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SETD6. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SETD6. Alternate names: FLJ21148ApplicationWB. Species reactivity: Human, Pig, Rabbit
Catalog No. 50-014-53872