Learn More
Eurofins Discoverx SETD7, 50 µg. (EPI056)
Supplier:  Eurofins Discoverx EPI056
Encompass
SETD7 is purified, C terminal 6xHis tagged recombinant SETD7 enzyme. This enzyme contains amino acids 186-352. It retains its binding/enzymatic activity and is intended for use in epigenetic assays. This enzyme is provided as 1 mg/mL frozen in 65% glycerol. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-200-5274