Learn More
HORIZON DISCOVERY BIOSCIENCES LIMITED SETDB1(HU) SIRNA ONTRGT+ 5NMOL
Catalog No. NC1860438
Encompass
Off-targets reduced by up to 90% compared to unmodified siRNAGuaranteed silencing by SMARTpool and 3 of 4 individual siRNAs Sequence information provided with siRNA purchase.DHARMACON mfg# L-020070-00-0005
Catalog No. NC1860438 Supplier HORIZON DISCOVERY BIOSCIENCES LIMITED Supplier No. L020070000005
Off-targets reduced by up to 90% compared to unmodified siRNAGuaranteed silencing by SMARTpool and 3 of 4 individual siRNAs Sequence information provided with siRNA purchase.DHARMACON mfg# L-020070-00-0005