Learn More
Aviva Systems Biology Corp SETDB2 Antibody - N-terminal region (ARP48955_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48955_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SETDB2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SETDB2. Alternate names: CLLD8, CLLL8, KMT1F, C13orf4ApplicationWB. Species reactivity: Guinea Pig, Human, Rat
Catalog No. 50-014-59974