Learn More
Cayman Chemical Sethyl N4 trIfluoromethyl 10mg
SDP
Catalog No. 502893464
Encompass_Preferred
EPIT is a selective, competitive inhibitor of nNOS having a Ki of 0.32 µM for the purified human enzyme. EPIT exhibits 115-fold and 29-fold selectivity for nNOS compared to iNOS and eNOS, respectively. It exhibits reduced inhibitory potency in whole-cell assays, possibly due to reduced intracellular uptake.{5065}
Catalog No. 50-289-3464 Supplier Cayman Chemical Supplier No. 8128010 MG
EPIT is a selective, competitive inhibitor of nNOS having a Ki of 0.32 µM for the purified human enzyme. EPIT exhibits 115-fold and 29-fold selectivity for nNOS compared to iNOS and eNOS, respectively. It exhibits reduced inhibitory potency in whole-cell assays, possibly due to reduced intracellular uptake.{5065}