Learn More
Selleck Chemical LLC Setipiprant
SDP
Catalog No. 5000003619
Encompass_Preferred
Setipiprant (ACT-129968 KYTH-105) is a selective orally available antagonist of the prostaglandin D2 receptor 2 (DP2) that that has been shown to have greater specificity for DP2 (CRTH2) than for DP1
Catalog No. 50-000-03619 Supplier Selleck Chemical LLC Supplier No. S575325MG
Setipiprant (ACT-129968 KYTH-105) is a selective orally available antagonist of the prostaglandin D2 receptor 2 (DP2) that that has been shown to have greater specificity for DP2 (CRTH2) than for DP1