Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Setiptiline maleate | 85650-57-3 | 99.9% | 377.43 | 1 ML
SDP

Catalog No. 5000402115
Encompass_Preferred

Setiptiline maleate is a tetracyclic antidepressant (TeCA) that functions as a noradrenergic and specific serotonergic antidepressant (NaSSA). It acts as a norepinephrine reuptake inhibitor and an α2-adrenergic receptor antagonist, as well as a serotonin receptor antagonist, likely affecting the 5-HT2A, 5-HT2C, and/or 5-HT3 subtypes. It also functions as an H1 receptor inverse agonist/antihistamine, making it suitable for research applications.

  • High purity (99.94%) for reliable research
  • Well-characterized pharmacological actions
  • Inhibits norepinephrine reuptake
  • Antagonizes α2-adrenergic receptors
  • Antagonizes serotonin receptors (5-HT2A, 5-HT2C, and/or 5-HT3)
  • Acts as an H1 receptor inverse agonist/antihistamine
  • Detailed solubility information and protocols for in vitro and in vivo studies

Catalog No. 50-004-02115 Supplier Medchemexpress LLC Supplier No. HY32329A10MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Setiptiline maleate is a tetracyclic antidepressant (TeCA) that functions as a noradrenergic and specific serotonergic antidepressant (NaSSA). It acts as a norepinephrine reuptake inhibitor and an α2-adrenergic receptor antagonist, as well as a serotonin receptor antagonist, likely affecting the 5-HT2A, 5-HT2C, and/or 5-HT3 subtypes. It also functions as an H1 receptor inverse agonist/antihistamine, making it suitable for research applications.

  • High purity (99.94%) for reliable research
  • Well-characterized pharmacological actions
  • Inhibits norepinephrine reuptake
  • Antagonizes α2-adrenergic receptors
  • Antagonizes serotonin receptors (5-HT2A, 5-HT2C, and/or 5-HT3)
  • Acts as an H1 receptor inverse agonist/antihistamine
  • Detailed solubility information and protocols for in vitro and in vivo studies

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.