Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Setiptiline (maleate) | 85650-57-3 | 99.9% | 377.43 | 100 MG
SDP

Catalog No. 5000400428
Encompass_Preferred

Setiptiline maleate is a serotonin receptor antagonist. It is classified as a tetracyclic antidepressant (TeCA) and acts as a noradrenergic and specific serotonergic antidepressant (NaSSA). It functions as a norepinephrine reuptake inhibitor, an α2-adrenergic receptor antagonist, and a serotonin receptor antagonist, potentially targeting the 5-HT2A, 5-HT2C, and/or 5-HT3 subtypes. Additionally, it acts as an H1 receptor inverse agonist/antihistamine. It is intended for research use only.

  • Serotonin receptor antagonist
  • Tetracyclic antidepressant (TeCA)
  • Noradrenergic and specific serotonergic antidepressant (NaSSA)
  • Norepinephrine reuptake inhibitor
  • Alpha2-adrenergic receptor antagonist
  • Antagonizes serotonin receptor subtypes: 5-HT2A, 5-HT2C, and/or 5-HT3
  • H1 receptor inverse agonist/antihistamine
  • Purity: 99.94%
  • Appearance: Solid
  • Color: White to gray

Catalog No. 50-004-00428 Supplier Medchemexpress LLC Supplier No. HY32329A100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Setiptiline maleate is a serotonin receptor antagonist. It is classified as a tetracyclic antidepressant (TeCA) and acts as a noradrenergic and specific serotonergic antidepressant (NaSSA). It functions as a norepinephrine reuptake inhibitor, an α2-adrenergic receptor antagonist, and a serotonin receptor antagonist, potentially targeting the 5-HT2A, 5-HT2C, and/or 5-HT3 subtypes. Additionally, it acts as an H1 receptor inverse agonist/antihistamine. It is intended for research use only.

  • Serotonin receptor antagonist
  • Tetracyclic antidepressant (TeCA)
  • Noradrenergic and specific serotonergic antidepressant (NaSSA)
  • Norepinephrine reuptake inhibitor
  • Alpha2-adrenergic receptor antagonist
  • Antagonizes serotonin receptor subtypes: 5-HT2A, 5-HT2C, and/or 5-HT3
  • H1 receptor inverse agonist/antihistamine
  • Purity: 99.94%
  • Appearance: Solid
  • Color: White to gray

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.