Learn More
Medchemexpress LLC Setiptiline maleate | 85650-57-3 | 99.9% | 377.43 | 50 MG
SDP
Catalog No. 5000401121
Encompass_Preferred
Setiptiline maleate is a serotonin receptor antagonist and a tetracyclic antidepressant (TeCA) that acts as a noradrenergic and specific serotonergic antidepressant (NaSSA).
  • Functions as a norepinephrine reuptake inhibitor.
  • Acts as an α2-adrenergic receptor antagonist.
  • Serves as a serotonin receptor antagonist, targeting 5-HT2A, 5-HT2C, and/or 5-HT3 subtypes.
  • Acts as an H1 receptor inverse agonist/antihistamine.
Catalog No. 50-004-01121 Supplier Medchemexpress LLC Supplier No. HY32329A50MG
Setiptiline maleate is a serotonin receptor antagonist and a tetracyclic antidepressant (TeCA) that acts as a noradrenergic and specific serotonergic antidepressant (NaSSA).
  • Functions as a norepinephrine reuptake inhibitor.
  • Acts as an α2-adrenergic receptor antagonist.
  • Serves as a serotonin receptor antagonist, targeting 5-HT2A, 5-HT2C, and/or 5-HT3 subtypes.
  • Acts as an H1 receptor inverse agonist/antihistamine.