Learn More
TARGETMOL CHEMICALS INC Setmelanotide Acetate 1MG
SDP
Catalog No. 502877508
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Setmelanotide Acetate(920014-72-8 free base) (RM-493 Acetate) is a selective agonist of melanocortin 4 receptor (MC4R)(human and rat MC4R with EC50s of 0.27 nM and 0.28 nM, respectively). Purity 100%
Catalog No. 50-287-7508 Supplier TARGETMOL CHEMICALS INC Supplier No. T12882L1MG
Also available in 1 mL, 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Setmelanotide Acetate(920014-72-8 free base) (RM-493 Acetate) is a selective agonist of melanocortin 4 receptor (MC4R)(human and rat MC4R with EC50s of 0.27 nM and 0.28 nM, respectively). Purity 100%