Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Setmelanotide monoacetate | 2759937-80-7 | 99.7% | 1177.36 g/mol | C51H72N18O11S2 | 10 MG
SDP

Catalog No. 5000297742
Encompass_Preferred

Setmelanotide monoacetate is a selective melanocortin 4 receptor (MC4R) agonist peptide intended for laboratory research. It is supplied as a white to off-white solid with supporting analytical data and storage guidance for both solution and bulk formats.

  • Potent MC4R agonist with EC50 ≈ 0.27 nM (human and rat).
  • Purity: 99.71% by HPLC.
  • Molecular weight: 1177.36 g/mol.
  • Chemical formula: C51H72N18O11S2.
  • Physical appearance: white to off-white solid.
  • Storage: store sealed at -20°C; in solvent -80°C up to 6 months, -20°C up to 1 month.
  • Available pack sizes: 1 mg, 5 mg, 10 mg, 25 mg; larger sizes available by quote.
  • Analytical documents available: data sheet, COA, SDS, HNMR, LCMS.

Catalog No. 50-002-97742 Supplier Medchemexpress LLC Supplier No. HY19870C10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Setmelanotide monoacetate is a selective melanocortin 4 receptor (MC4R) agonist peptide intended for laboratory research. It is supplied as a white to off-white solid with supporting analytical data and storage guidance for both solution and bulk formats.

  • Potent MC4R agonist with EC50 ≈ 0.27 nM (human and rat).
  • Purity: 99.71% by HPLC.
  • Molecular weight: 1177.36 g/mol.
  • Chemical formula: C51H72N18O11S2.
  • Physical appearance: white to off-white solid.
  • Storage: store sealed at -20°C; in solvent -80°C up to 6 months, -20°C up to 1 month.
  • Available pack sizes: 1 mg, 5 mg, 10 mg, 25 mg; larger sizes available by quote.
  • Analytical documents available: data sheet, COA, SDS, HNMR, LCMS.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.