Learn More
Medchemexpress LLC Setmelanotide (monoacetate) | 2759937-80-7 | 99.8% | 1177.36 | 25 MG
SDP
Catalog No. 5000402705
Encompass_Preferred
Setmelanotide (monoacetate) is a melanocortin 4 receptor (MC4R) agonist. It acts on human and rat MC4R with EC50 values of 0.27 nM and 0.28 nM, respectively, and is intended for research use only.
  • Acts on human and rat MC4R
  • Stable as a solid
  • Suitable for research use
Catalog No. 50-004-02705 Supplier Medchemexpress LLC Supplier No. 25MGHY19870C
Setmelanotide (monoacetate) is a melanocortin 4 receptor (MC4R) agonist. It acts on human and rat MC4R with EC50 values of 0.27 nM and 0.28 nM, respectively, and is intended for research use only.
  • Acts on human and rat MC4R
  • Stable as a solid
  • Suitable for research use