Learn More
Medchemexpress LLC Setmelanotide monoacetate | 2759937-80-7 | 99.9% | 1177.36 | 5 MG
SDP
Catalog No. 5000401489
Encompass_Preferred
Setmelanotide (monoacetate) is a melanocortin 4 receptor (MC4R) agonist that acts on human and rat MC4R with EC50 values of 0.27 nM and 0.28 nM, respectively. It is for research use only and not sold to patients.
  • Melanocortin 4 receptor (MC4R) agonist
  • Acts on human and rat MC4R
  • EC50 values of 0.27 nM (human) and 0.28 nM (rat)
  • Available for bulk orders
  • Supported by 8 publications in Google Scholar
Catalog No. 50-004-01489 Supplier Medchemexpress LLC Supplier No. HY19870C5MG
Setmelanotide (monoacetate) is a melanocortin 4 receptor (MC4R) agonist that acts on human and rat MC4R with EC50 values of 0.27 nM and 0.28 nM, respectively. It is for research use only and not sold to patients.
  • Melanocortin 4 receptor (MC4R) agonist
  • Acts on human and rat MC4R
  • EC50 values of 0.27 nM (human) and 0.28 nM (rat)
  • Available for bulk orders
  • Supported by 8 publications in Google Scholar