Learn More
Selleck Chemical LLC Setmelanotide-E1002-5MG
SDP
Supplier:  Selleck Chemical LLC E10025MG
Encompass_Preferred
Setmelanotide (RM-493 BIM-22493 CAM 4072) is a cyclic peptide full agonist of melanocortin-4 receptor (MC4R) with an EC50 of 0 27 nM and a Ki of 2 1 nM
Catalog No. 50-239-0033