Learn More
Med Vet International Vacutainer Blood Collection Set 23g, x 3/4in, Winged, Green, Safety, 50/Bx
Catalog No. 503043033
Encompass_Preferred
Pushbutton safety mechanism is activated while still in the vein to help protect against needlestick injury 1-handed activation leaves a hand free for patient care Ideal for high-risk environments
Catalog No. 50-304-3033 Supplier Med Vet International Supplier No. 367342
Pushbutton safety mechanism is activated while still in the vein to help protect against needlestick injury 1-handed activation leaves a hand free for patient care Ideal for high-risk environments