Learn More
MSC SETU
Catalog No. NC3359008
Encompass
Setup Block Sets; Type: Rectangular; Squareness Per 1 Inch (Decimal Inch): 0.001; Overall Length (Inch): 3; Number of Pieces: 15; Includes: Fitted Case
Catalog No. NC3359008 Supplier MSC Supplier No. 17621533
Setup Block Sets; Type: Rectangular; Squareness Per 1 Inch (Decimal Inch): 0.001; Overall Length (Inch): 3; Number of Pieces: 15; Includes: Fitted Case