Learn More
Medchemexpress LLC Sevelamer (carbonate) | 845273-93-0 | 25 G
SDP
Catalog No. 5000349402
Encompass_Preferred
Sevelamer carbonate is an orally active and non-calcium-based phosphate binding agent used for research on hyperphosphatemia of chronic kidney disease (CKD). It effectively lowers serum phosphorus levels with minimal effect on serum calcium or serum chloride levels in vivo. It is considered an improved, buffered form of sevelamer. This product is intended for laboratory use in the manufacture of substances and is not classified as a hazardous substance or mixture.
  • Orally active and non-calcium-based phosphate binding agent
  • Used for research on hyperphosphatemia of chronic kidney disease (CKD)
  • Effectively lowers serum phosphorus levels
  • Minimal effect on serum calcium or serum chloride levels in vivo
  • Stable under recommended storage conditions
  • For research use only
Catalog No. 50-003-49402 Supplier Medchemexpress LLC Supplier No. 25GHY13995B
Sevelamer carbonate is an orally active and non-calcium-based phosphate binding agent used for research on hyperphosphatemia of chronic kidney disease (CKD). It effectively lowers serum phosphorus levels with minimal effect on serum calcium or serum chloride levels in vivo. It is considered an improved, buffered form of sevelamer. This product is intended for laboratory use in the manufacture of substances and is not classified as a hazardous substance or mixture.
  • Orally active and non-calcium-based phosphate binding agent
  • Used for research on hyperphosphatemia of chronic kidney disease (CKD)
  • Effectively lowers serum phosphorus levels
  • Minimal effect on serum calcium or serum chloride levels in vivo
  • Stable under recommended storage conditions
  • For research use only