Learn More
Medchemexpress LLC Sevelamer carbonate | 845273-93-0 | MFCD18904375 | 5 G
SDP
Catalog No. 5000352092
Encompass_Preferred
Sevelamer carbonate is an orally active, non-calcium-based phosphate binding agent used in research for hyperphosphatemia of chronic kidney disease (CKD). It effectively lowers serum phosphorus levels with minimal impact on serum calcium or chloride levels in vivo. It is considered an improved, buffered form of sevelamer, with carbonate replacing chloride as the anion.
  • Lowers serum phosphorus levels
  • May cause less metabolic acidosis
  • Effective in attenuating chronic hyperphosphatemia in animal models
Catalog No. 50-003-52092 Supplier Medchemexpress LLC Supplier No. HY13995B5G
Sevelamer carbonate is an orally active, non-calcium-based phosphate binding agent used in research for hyperphosphatemia of chronic kidney disease (CKD). It effectively lowers serum phosphorus levels with minimal impact on serum calcium or chloride levels in vivo. It is considered an improved, buffered form of sevelamer, with carbonate replacing chloride as the anion.
  • Lowers serum phosphorus levels
  • May cause less metabolic acidosis
  • Effective in attenuating chronic hyperphosphatemia in animal models