Learn More
Medchemexpress LLC Sevelamer (hydrochloride) | 152751-57-0 | 98.0% | 186.08 (monomer) | 1 G
SDP
Catalog No. 5000352437
Encompass_Preferred
Sevelamer (hydrochloride) is an orally active phosphate binding agent primarily used for research into hyperphosphatemia associated with chronic kidney disease. This substance consists of polyallylamine crosslinked with epichlorohydrin. It has a molecular weight of 186.08 (monomer) and is stable under recommended storage conditions, appearing as a white to off-white solid. It is classified as harmful to aquatic life with long lasting effects.
  • Orally active phosphate binding agent
  • Used for research of hyperphosphatemia with chronic kidney disease
  • Decreases serum levels of gut-derived uremic toxins
  • Reduces serum phosphate levels
  • Stable under recommended storage conditions
  • Solid appearance
Catalog No. 50-003-52437 Supplier Medchemexpress LLC Supplier No. HY13995A1G
Sevelamer (hydrochloride) is an orally active phosphate binding agent primarily used for research into hyperphosphatemia associated with chronic kidney disease. This substance consists of polyallylamine crosslinked with epichlorohydrin. It has a molecular weight of 186.08 (monomer) and is stable under recommended storage conditions, appearing as a white to off-white solid. It is classified as harmful to aquatic life with long lasting effects.
  • Orally active phosphate binding agent
  • Used for research of hyperphosphatemia with chronic kidney disease
  • Decreases serum levels of gut-derived uremic toxins
  • Reduces serum phosphate levels
  • Stable under recommended storage conditions
  • Solid appearance