Learn More
Medchemexpress LLC Sevelamer hydrochloride | 152751-57-0 | 98.0% | 186.08 (monomer) | 500 MG
SDP
Catalog No. 5000351284
Encompass_Preferred
Sevelamer hydrochloride is an orally active phosphate binding agent used for research of hyperphosphatemia with chronic kidney disease. It consists of polyallylamine crosslinked with epichlorohydrin.
Catalog No. 50-003-51284 Supplier Medchemexpress LLC Supplier No. HY13995A500MG
Sevelamer hydrochloride is an orally active phosphate binding agent used for research of hyperphosphatemia with chronic kidney disease. It consists of polyallylamine crosslinked with epichlorohydrin.