Learn More
Medchemexpress LLC HY-15996A 5mg Medchemexpress, Seviteronel (R enantiomer) CAS:1375603-38-5 Purity:>98%
SDP
Catalog No. 502081858
Encompass_Preferred
Medchemexpress, HY-15996A 5mg Seviteronel (R enantiomer) CAS:1375603-38-5 Seviteronel R enantiomer (VT-464 R enantiomer) is the R enantiomer of Seviteronel (VT-464), which is a potent CYP17 lyase inhibitor(h-Lyase IC50=69 nM); Seviteronel (VT-464) R enantiomer's activity is unknown. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1858 Supplier Medchemexpress LLC Supplier No. HY15996A5MG
Medchemexpress, HY-15996A 5mg Seviteronel (R enantiomer) CAS:1375603-38-5 Seviteronel R enantiomer (VT-464 R enantiomer) is the R enantiomer of Seviteronel (VT-464), which is a potent CYP17 lyase inhibitor(h-Lyase IC50=69 nM); Seviteronel (VT-464) R enantiomer's activity is unknown. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.