Learn More
TARGETMOL CHEMICALS INC Seviteronel R enantiomer 1MG
SDP
Catalog No. 502879622
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Seviteronel R enantiomer (VT-464 R enantiomer) is a Seviteronel isomer produced during the manufacturing process. Purity 99.98%
Catalog No. 50-287-9622 Supplier TARGETMOL CHEMICALS INC Supplier No. T133121MG
Also available in 1 mL, 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Seviteronel R enantiomer (VT-464 R enantiomer) is a Seviteronel isomer produced during the manufacturing process. Purity 99.98%