Learn More
Medchemexpress LLC HY-15996B 5mg Medchemexpress, Seviteronel (racemate) CAS:1375603-36-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY15996B5MG
Encompass_Preferred
Medchemexpress, HY-15996B 5mg Seviteronel (racemate) CAS:1375603-36-3 Seviteronel racemate (VT-464 racemate) is the racemate form of Seviteronel (VT-464), which is a potent CYP17 lyase inhibitor(h-Lyase IC50=nM)inhibition. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1859