Learn More
Med Vet International Sevoflurane Anti-Spill Pour Spout, J0562Y
Catalog No. 5000585233
Encompass_Preferred
This handy adapter screws onto gas agent bottle top andthe other end fits snugly into the filling funnel on the gas vaporizer. It makes vaporizerrefilling spill free. Color coded for agent type.
Catalog No. 50-005-85233 Supplier Med Vet International Supplier No. JOR562Y
This handy adapter screws onto gas agent bottle top andthe other end fits snugly into the filling funnel on the gas vaporizer. It makes vaporizerrefilling spill free. Color coded for agent type.