Learn More
Medchemexpress LLC SEW2871 | 256414-75-2 | 99.9% | 440.36 | 10 MG
SDP
Catalog No. 5000453596
Encompass_Preferred
SEW2871 is an orally active, potent, highly selective S1P1 (sphingosine-1-phosphate type 1 receptor) agonist, with an EC50 of 13.8 nM. It activates ERK, Akt, and Rac signaling pathways and induces S1P1 internalization and recycling. SEW2871 reduces lymphocyte numbers in blood and can be used for the research of diabetes, Alzheimer's disease, liver fibrosis, and inflammatory responses.
  • Orally active, potent, highly selective S1P1 agonist.
  • Activates ERK, Akt, and Rac signaling pathways.
  • Induces S1P1 internalization and recycling.
  • Reduces lymphocyte numbers in blood.
  • Can be used for research of diabetes, Alzheimer's disease, liver fibrosis, and inflammatory responses.
Catalog No. 50-004-53596 Supplier Medchemexpress LLC Supplier No. HYW00894710MG
SEW2871 is an orally active, potent, highly selective S1P1 (sphingosine-1-phosphate type 1 receptor) agonist, with an EC50 of 13.8 nM. It activates ERK, Akt, and Rac signaling pathways and induces S1P1 internalization and recycling. SEW2871 reduces lymphocyte numbers in blood and can be used for the research of diabetes, Alzheimer's disease, liver fibrosis, and inflammatory responses.
  • Orally active, potent, highly selective S1P1 agonist.
  • Activates ERK, Akt, and Rac signaling pathways.
  • Induces S1P1 internalization and recycling.
  • Reduces lymphocyte numbers in blood.
  • Can be used for research of diabetes, Alzheimer's disease, liver fibrosis, and inflammatory responses.