Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Seward™ Stomacher™ Simplette™ Pipets

Catalog No. 142856
Change view
Click to view available options
Packaging Quantity:
2000/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging Quantity
14-285-6 2000/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-285-6 Supplier Seward™ Supplier No. 030020359
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Seward Simplettes™ provide an accurate and economical alternative to pipette tips, designed specifically to pipette samples from the bottom of the Stomacher™ bag without risk of cross contamination.

Helps facilitate sample transfer; Are of the right length to reach down to the bottom of the bag to recover the sample for inoculating Petri dishes and broths; Come in 2 sizes standard and long; Capable of pipetting volumes between 0.1ml and 2ml; irradiated sterile

Specifications

Product Type Transfer Pipet
Diameter (Metric) 4.1 mm
Length (Metric) 177 mm
Description Simplettes; Transfer Pipet
For Use With (Application) Ideal to pipette samples from sample bag
For Use With (Equipment) All Stomacher sample bags
Packaging Quantity 2000/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.