Learn More
MSC Value Collection Sewer Hose, -40 to 160 °F, 4,400 psi, 1/4 Inch ID, Black
Catalog No. 502835852
Encompass
This sewer hose is designed for demanding high-pressure applications, offering reliable performance across a wide temperature range. It is suitable for various industrial and commercial uses requiring a robust and durable hose.
  • Withstands temperatures from -40 to 160 °F.
  • Handles maximum working pressure up to 4,400 psi.
  • Features a 1/4 inch inside diameter.
  • Comes in a black finish.
  • Manufactured in Italy, ensuring quality construction.
  • Supplied as a 100 foot roll.
Catalog No. 50-283-5852 Supplier MSC Supplier No. 59814392
This sewer hose is designed for demanding high-pressure applications, offering reliable performance across a wide temperature range. It is suitable for various industrial and commercial uses requiring a robust and durable hose.
  • Withstands temperatures from -40 to 160 °F.
  • Handles maximum working pressure up to 4,400 psi.
  • Features a 1/4 inch inside diameter.
  • Comes in a black finish.
  • Manufactured in Italy, ensuring quality construction.
  • Supplied as a 100 foot roll.