Learn More
New England Biolabs, Inc. SexAI – 1000 units
SDP
Catalog No. 50812331
Encompass
A restriction endonuclease that recognizes the sequence A^CCWGG_T.
Catalog No. 50-812-331 Supplier New England Biolabs, Inc. Supplier No. R0605L
A restriction endonuclease that recognizes the sequence A^CCWGG_T.