Learn More
New England Biolabs, Inc. SexAI – 200 units
SDP
Catalog No. 50812332
Encompass
A restriction endonuclease that recognizes the sequence A^CCWGG_T.
Catalog No. 50-812-332 Supplier New England Biolabs, Inc. Supplier No. R0605S
A restriction endonuclease that recognizes the sequence A^CCWGG_T.