Learn More
Medchemexpress LLC SEZ6/BSRP-C Mouse 50ug | 50ug
SDP
Catalog No. 5000187681
Encompass_Preferred
The SEZ6/BSRP-C protein contributes significantly to cell-to-cell recognition and neuronal membrane signaling playing a crucial role in fundamental processes of neural networks SEZ6/BSRP-C is particularly important in balancing dendritic elongation and branching which shapes the complex structure of neurons SEZ6/BSRP-C Protein Mouse (HEK293 His) is the recombinant mouse-derived SEZ6/BSRP-C protein expressed by HEK293 with C-His labeled tag
Catalog No. 50-001-87681 Supplier Medchemexpress LLC Supplier No. HYP7803750UG
The SEZ6/BSRP-C protein contributes significantly to cell-to-cell recognition and neuronal membrane signaling playing a crucial role in fundamental processes of neural networks SEZ6/BSRP-C is particularly important in balancing dendritic elongation and branching which shapes the complex structure of neurons SEZ6/BSRP-C Protein Mouse (HEK293 His) is the recombinant mouse-derived SEZ6/BSRP-C protein expressed by HEK293 with C-His labeled tag