Learn More
Medchemexpress LLC SEZ6L2/BSRP-A Human 10ug | 10ug
SDP
Catalog No. 5000186256
Encompass_Preferred
SEZ6L2/BSRP-A potentially contributing to specialized functions in the endoplasmic reticulum (ER) of neurons hints at a role critical for neuronal homeostasis Its significance in the intricate molecular processes essential for neuronal function underscores the need for further exploration to unveil specific mechanisms and pathways through which SEZ6L2/BSRP-A participates in specialized ER functions SEZ6L2/BSRP-A Protein Human (HEK293 His) is the recombinant human-derived SEZ6L2/BSRP-A protein expressed by HEK293 with C-His labeled tag
Catalog No. 50-001-86256 Supplier Medchemexpress LLC Supplier No. HYP7720010UG
SEZ6L2/BSRP-A potentially contributing to specialized functions in the endoplasmic reticulum (ER) of neurons hints at a role critical for neuronal homeostasis Its significance in the intricate molecular processes essential for neuronal function underscores the need for further exploration to unveil specific mechanisms and pathways through which SEZ6L2/BSRP-A participates in specialized ER functions SEZ6L2/BSRP-A Protein Human (HEK293 His) is the recombinant human-derived SEZ6L2/BSRP-A protein expressed by HEK293 with C-His labeled tag