Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lonza Walkersville SF Cell Line 4D-Nucleofector LV Kit XL, Catalog Number V4LC-2002, Reaction Volume Up to 20 mL, Kit Size 5 reactions, Processing Time Less than 10 min, Compatible System 4D-Nucleofector LV Unit

Catalog No. NC2009675
Encompass

The Lonza SF Cell Line 4D-Nucleofector LV Kit XL is specifically designed for efficient, large-scale transfection of hard-to-transfect cell lines using the 4D-Nucleofector LV Unit. This closed-system nucleofection kit modifies up to 1 x 10^9 cells effectively, retaining cell viability. It allows variable reaction volumes and rapid processing, making it ideal for transient protein production and generating modified cell lines for various assays.

  • Compatible system: 4D-Nucleofector LV Unit
  • Reaction volume: Up to 20 mL
  • Cell numbers supported: Up to 1 x 10^9 cells
  • Processing time: Less than 10 minutes
  • Kit size: 5 reactions
  • Optimal cell lines: HEK293 suspension clones, CHO-S suspension clones, K-562, U-937
  • Applications: Large scale transient protein production, generation of modified cell lines
  • Key benefits: High transfection efficiencies, maintained cell functionality
  • Storage conditions: 2 to 8 °C

Catalog No. NC2009675 Supplier Lonza Walkersville Supplier No. V4LC2002
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Lonza SF Cell Line 4D-Nucleofector LV Kit XL is specifically designed for efficient, large-scale transfection of hard-to-transfect cell lines using the 4D-Nucleofector LV Unit. This closed-system nucleofection kit modifies up to 1 x 10^9 cells effectively, retaining cell viability. It allows variable reaction volumes and rapid processing, making it ideal for transient protein production and generating modified cell lines for various assays.

  • Compatible system: 4D-Nucleofector LV Unit
  • Reaction volume: Up to 20 mL
  • Cell numbers supported: Up to 1 x 10^9 cells
  • Processing time: Less than 10 minutes
  • Kit size: 5 reactions
  • Optimal cell lines: HEK293 suspension clones, CHO-S suspension clones, K-562, U-937
  • Applications: Large scale transient protein production, generation of modified cell lines
  • Key benefits: High transfection efficiencies, maintained cell functionality
  • Storage conditions: 2 to 8 °C

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.