Learn More
Lonza Walkersville SF Cell Line 96-well Nucleofector Kit (960 RCT)
Supplier:  Lonza Walkersville V4SC2960
Encompass
The SF Cell Line 96-well Nucleofector Kit is one of our three kits suited for medium-throughput transfection of cell lines when working with the 4D-Nucleofector 96-well Unit.Benefits:High transfection efficiencies in difficult-to-transfect cell linesCompatible with liquid handling systems for automated high-throughput electroporationConditions are transferable between 4D-Nucleofector System, 4D-Nucleofector 96-well Unit and 384-well Nucleofector SystemApplications:Transfection of low cell numbers in the range of 5 x 10⁴ to 1 x 10⁶ cellsHigh-throughput applications, e.g. screens of arrayed cDNA, RNAi or CRISPR librariesContent:22.5 mL SF Cell Line Nucleofector Solution5 mL Supplement 150 µg pmaxGFP Vector (1 μg/μl in 10 mM Tris pH 8.0)10 x 96-well Nucleocuvette Plate (20 μL)
Catalog No. NC1986639
Only null left
Add to Cart
Request a Quote