Learn More
IDEX Health & Science Upchurch Super Flangeless Ferrule, 6-32 or 6-40 Flat-Bottom, for 1/16" OD Tubing Natural, 10/pk
Catalog No. NC1819661
Encompass_Preferred
Super Flangeless Ferrules eliminate loosening of fittings due to tube twisting, hold tight even during vibration and delivers the highest pressure holding for flat bottom fittings that we offer. It is for a 1/16"OD tubing and a 6-32 or 6-40 Flat-Bottom port. The part includes the PEEK ferrule and stainless steel locking ring.
Catalog No. NC1819661 Supplier IDEX Health & Science Upchurch Supplier No. M650X
Super Flangeless Ferrules eliminate loosening of fittings due to tube twisting, hold tight even during vibration and delivers the highest pressure holding for flat bottom fittings that we offer. It is for a 1/16"OD tubing and a 6-32 or 6-40 Flat-Bottom port. The part includes the PEEK ferrule and stainless steel locking ring.