Learn More
Apexbio Technology LLC Serum-Free Cell Freezing Medium (HEK293 cells) 100ml
SDP
Catalog No. 5000063450
Encompass_Preferred
Serum-Free Cell Freezing Medium for HEK293 cells is developed for long-term preservation of HEK293 suspension cell lines in liquid nitrogen storage This ready-to-use freezing solution contains defined chemical composition including essential nutrients (glucose amino acids) cryoprotectant DMSO and phenol red It is serum-free and devoid of animal-origin proteins thereby reducing risks associated with serum variability and foreign protein contamination during cell culture Upon thawing HEK293 cells cryopreserved with this reagent typically exhibit a recovery rate higher than 85% This formulation is suitable for routine freezing and revival of HEK293 cells widely utilized in research applications such as recombinant protein expression viral vector production and gene-function analysis
Catalog No. 50-000-63450 Supplier Apexbio Technology LLC Supplier No. K1148100
Add to Cart
Add to Cart
Serum-Free Cell Freezing Medium for HEK293 cells is developed for long-term preservation of HEK293 suspension cell lines in liquid nitrogen storage This ready-to-use freezing solution contains defined chemical composition including essential nutrients (glucose amino acids) cryoprotectant DMSO and phenol red It is serum-free and devoid of animal-origin proteins thereby reducing risks associated with serum variability and foreign protein contamination during cell culture Upon thawing HEK293 cells cryopreserved with this reagent typically exhibit a recovery rate higher than 85% This formulation is suitable for routine freezing and revival of HEK293 cells widely utilized in research applications such as recombinant protein expression viral vector production and gene-function analysis