Learn More
Kuhner Shaker Inc SF-X SHAKING UNIT..CONTROLLED
Catalog No. 502570046
Encompass
SF-X Shaking unit..Controlled acceleration rate included..User Selectable Orbit (12.5mm, 25mm, 50mm, and User Specified)..Loads up to 25kg / shaker (>25kg possible)..Direct drive with single eccentric (no belt)..100% stainless steel interior..
Catalog No. 50-257-0046 Supplier Kuhner Shaker Inc Supplier No. SMX1610
Add to Cart
Add to Cart
Request a Quote
SF-X Shaking unit..Controlled acceleration rate included..User Selectable Orbit (12.5mm, 25mm, 50mm, and User Specified)..Loads up to 25kg / shaker (>25kg possible)..Direct drive with single eccentric (no belt)..100% stainless steel interior..