Learn More
Aviva Systems Biology Corp SF1 Antibody - N-terminal region (ARP41214_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41214_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against SF1. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human SF1. Alternate names: BBP, MBBP, ZFM1, ZNF162, D11S636ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-57766