Learn More
Medchemexpress LLC HY-15842 5mg Medchemexpress, SF1670 CAS:345630-40-2 Purity:>98%
SDP
Catalog No. 502030690
Encompass_Preferred
Medchemexpress, HY-15842 5mg SF1670 CAS:345630-40-2 SF1670 is a potent and specific phosphatase and tensin homolog deleted on chromosome 10 (PTEN) inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0690 Supplier Medchemexpress LLC Supplier No. HY158425MG
Medchemexpress, HY-15842 5mg SF1670 CAS:345630-40-2 SF1670 is a potent and specific phosphatase and tensin homolog deleted on chromosome 10 (PTEN) inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.