Learn More
Medchemexpress LLC SF2312 ammonium | 98.0% | 214.11 | 1 MG
SDP
Catalog No. 5000437325
Encompass_Preferred
SF2312 ammonium is the ammonium form of SF2312. SF2312 is a phosphonate antibiotic and enolase inhibitor. It can be isolated from the actinomycete Micromonospora and is active under anaerobic conditions. This product is selectively toxic to ENO1-deleted glioma cells.
  • Phosphonate antibiotic.
  • Enolase inhibitor.
  • Isolated from the actinomycete Micromonospora.
  • Active under anaerobic conditions.
  • Selectively toxic to ENO1-deleted glioma cells.
Catalog No. 50-004-37325 Supplier Medchemexpress LLC Supplier No. HY117778B1MG
SF2312 ammonium is the ammonium form of SF2312. SF2312 is a phosphonate antibiotic and enolase inhibitor. It can be isolated from the actinomycete Micromonospora and is active under anaerobic conditions. This product is selectively toxic to ENO1-deleted glioma cells.
  • Phosphonate antibiotic.
  • Enolase inhibitor.
  • Isolated from the actinomycete Micromonospora.
  • Active under anaerobic conditions.
  • Selectively toxic to ENO1-deleted glioma cells.