Learn More
Medchemexpress LLC SF2523 | 1174428-47-7 | 99.3% | 371.41 g/mol | C19H17NO5S | 5 MG
SDP
Catalog No. 501932053
Encompass_Preferred
SF2523 is a research-grade small-molecule inhibitor that targets PI3K family members and the bromodomain protein BRD4. It shows potent biochemical and cellular activity, decreasing MYCN and Cyclin D1 levels and inhibiting AKT signaling. Supplied as a solid or as a DMSO solution for preclinical research use.
  • Potent inhibition of PI3Kα, PI3Kγ, DNA-PK, BRD4, and mTOR (reported IC50s: 34 nM, 158 nM, 9 nM, 241 nM, 280 nM).
  • High purity (>99%).
  • Available in small solid sizes and as pre-made DMSO solutions.
  • Reduces MYCN and Cyclin D1 protein levels in cell-based assays.
  • Suitable for pathway inhibition studies of PI3K/AKT/mTOR and epigenetic regulation.
Catalog No. 50-193-2053 Supplier Medchemexpress LLC Supplier No. HY1011465MG
SF2523 is a research-grade small-molecule inhibitor that targets PI3K family members and the bromodomain protein BRD4. It shows potent biochemical and cellular activity, decreasing MYCN and Cyclin D1 levels and inhibiting AKT signaling. Supplied as a solid or as a DMSO solution for preclinical research use.
  • Potent inhibition of PI3Kα, PI3Kγ, DNA-PK, BRD4, and mTOR (reported IC50s: 34 nM, 158 nM, 9 nM, 241 nM, 280 nM).
  • High purity (>99%).
  • Available in small solid sizes and as pre-made DMSO solutions.
  • Reduces MYCN and Cyclin D1 protein levels in cell-based assays.
  • Suitable for pathway inhibition studies of PI3K/AKT/mTOR and epigenetic regulation.