Learn More
Aviva Systems Biology Corp SF3A3 Antibody - middle region (ARP40684_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40684_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SF3A3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SF3A3. Alternate names: PRP9, PRPF9, SAP61, SF3a60ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-57587