Learn More
Aviva Systems Biology Corp SF3B1 Antibody - N-terminal region (ARP40732_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40732_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SF3B1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SF3B1. Alternate names: PRP10, PRPF10, SAP155, SF3b155, MDS, Hsh155ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-53230