Learn More
Aviva Systems Biology Corp SF3B6 Antibody - middle region (ARP40829_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40829_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SF3B14. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SF3B14. Alternate names: CGI-110, HSPC175, Ht006, P14, SAP14, SF3B14aApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-53243