Learn More
Aviva Systems Biology Corp SF3B3 Antibody - middle region (ARP40730_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40730_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SF3B3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SF3B3. Alternate names: KIAA0017, RSE1, SAP130, SF3b130, STAF130ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-63012