Learn More
Aviva Systems Biology Corp SF3B4 Antibody - N-terminal region (ARP40610_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40610_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SF3B4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SF3B4. Alternate names: AFD1, Hsh49, SAP49, SF3b49ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-53213