Learn More
ABclonal Technology SF3B6 Rabbit pAb
SDP
Supplier:  ABclonal Technology A13097100UL
Encompass_Preferred
This gene encodes a 14 kDa protein subunit of the splicing factor 3b complex. Splicing factor 3b associates with both the U2 and U11/U12 small nuclear ribonucleoprotein complexes (U2 snRNP) of spliceosomes. This 14 kDa protein interacts directly with subunit 1 of the splicing factor 3b complex. This 14 kDa protein also interacts directly with the adenosine that carries out the first transesterification step of splicing at the pre-mRNA branch site.
Catalog No. 50-001-16482