Learn More
Aviva Systems Biology Corp SF4 Antibody - N-terminal region (ARP40961_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40961_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SF4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SF4. Alternate names: DKFZp434E2216, F23858, RBP, SF4ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat, Yeast, Zebrafish
Catalog No. 50-014-57676